Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Array Rat Phosphodiesterase Panel

Catalog No. 4378706
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-787-06 4 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-787-06 Supplier Applied Biosystems™ Supplier No. 4378706
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Applied Biosystems™ TaqMan™ Rat Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.

This product is designed with 48 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored (at 4°C.)
Form Dried Down
Format 384-well microfluidic card
Reaction Speed Standard
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Research Category Cell Cycle & Proliferation, Oncology, Toxicology & Drug Metabolism, Neurobiology, Inflammation
Species Rat
Target Includes five distinct phosphodiesterase families
For Use With (Application) Gene Expression
No. of Reactions 1 x 2 μL Reactions
Show More Show Less
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.