Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ GAPDH Assay, JUN™ dye/QSY™ probe, primer-limited
GREENER_CHOICE

Catalog No. 4485713
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-857-13 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-857-13 Supplier Applied Biosystems™ Supplier No. 4485713
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

Provides a pre-formulated assay for quantitating human GAPDH.

This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. It consists of a JUN™ dye-labeled QSY™ probe plus sequence-specific forward and reverse primers. The assay can be used for multiplex or singleplex PCR reactions, and it is primer limited, so it can be used with samples that overexpress GAPDH.

Save time in assay design, formulation, and testing by using the TaqMan™ GAPDH Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.

In order to facilitate multiplex experiments, the assay formulation is primer limited, allowing the assay to be used with high GAPDH expressors.

The JUN™ dye is optimized for use with the 4th filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments. Please note that master mix containing ROX™ dye cannot be used with this assay (with JUN™ dye) due to an overlap of spectra. TaqMan™ Multiplex Master Mix with MUSTANG PURPLE™ dye is recommended.

  • Eliminate assay design time by using pre-designed primers and pre-designed JUN™ dye-labeled TaqMan™ probe
  • Minimize experimental optimization time
  • Use in conjunction with FAM™ or VIC™ dye-labeled predesigned gene expression assays
  • Formulated at a primer-limited concentration for use in multiplex reactions
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_002046.3
Amplicon Size 118
Content And Storage Primer-Probe Set(s)
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Gene Symbol GAPDH
Internal Probe Modification QSY™ Non-Fluorescent Quencher (3')
Label or Dye JUN
Product Line TaqMan
Product Type Gapdh Assay
Quantity 250 reactions
Species Human
Sufficient For 250 Reactions
Target Glyceraldehyde-3-phosphate dehydrogenase
For Use With (Application) Gene Expression
No. of Reactions 250 Reactions
Show More Show Less
What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?

ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.

Please refer to your instrument user guide for additional information on compatibility with different dye channels.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.