Learn More
Applied Biosystems™ TaqMan™ Gene Expression Assay, VIC primer-limited

Description
Applied Biosystems TaqMan Gene Expression Assays, VIC primer-limited, are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5' end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3' end. These assays are primer-limited and ideal for multiplex reactions looking at two different gene targets in the same qPCR well. Note that for digital PCR, primer-limiting is typically not recommended. The full-concentration TaqMan assay (Cat. No. 4448491) is suggested if you are using dPCR.
Features include:
- Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
- Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
- Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
- Accurate—identify small fold-changes with high accuracy of quantitation
- Extensive content—over 1.8 million predesigned assays available for over 25 different species
- Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
- Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same qPCR well
- Ideal for control assays—use a VIC primer-limited assay for high-expressing endogenous control genes
Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe
Pre-formulated assay
- 2 unlabeled PCR primers (150 nM each final 1X concentration)
- 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)
TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents. All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays Guarantee.*
Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556)
*Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.
Specifications
Specifications
| Concentration | 20X |
| Content And Storage | 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Reaction Speed | Fast |
| For Use With (Equipment) | 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex |
| Product Line | TaqMan |
| Product Type | Gene Expression Assay |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.