Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TaqMan™ miRNA ABC Purification Kit - Human Panel A
GREENER_CHOICE

Catalog No. 501148000
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8000 40 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-8000 Supplier LIFE TECHNOLOGIES Supplier No. 4473087
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Content And Storage Buffer Kit - Store at room temperature
Lysis Buffer
ABC Buffer
Wash Buffer 1
Wash Buffer 2
Elution Buffer
1.5 mL LoBind tubes

Human Panel A Beads
Store at 2 °C to 8 °C
Format Kit
Isolation Technology Magnetic Bead
Purification Time 75 min.
RNAi Type miRNA
Species Human
Sample Type Cell Cultures, Plasma, Saliva, Serum, Tissue, Tissue (FFPE), Tissue Sections, Urine, Whole Blood
Final Product Type micro RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), microRNA Analysis
Product Line TaqMan
Product Type miRNA ABC Purification Kit
Quantity 40 Preps
Target miRNA
System Type Dynabeads™
Show More Show Less
I am using the TaqMan miRNA ABC Purification Kit and am having difficulty detecting miRNA in qPCR assays. What is the issue?

Here are possible causes and solutions:

- RNA degradation due to RNAse contamination during procedure: See “Guidelines to prevent RNAse contamination” on page 12 of the manual (https://tools.thermofisher.com/content/sfs/manuals/4473439C.pdf) for proper handling of sample.
- Inefficient elution of miRNA from the beads: follow hybridization, wash, and elution steps on pages 17-18.
- Inefficient washing: fully resuspend beads in wash buffers, and completely remove buffers at the end of the washes.
- Beads dried out during procedure: prepare the beads just before use and keep the beads suspended in liquid and the tube cap closed in between steps.
- Buffers not diluted as directed: prepare buffers as directed on page 13.
- Kit components compromised due to inappropriate storage conditions: store reagents as directed in “Kit contents and storage conditions” on page 9 of the manual.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.