Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Mutation Detection Assay with castPCR™ Technology
GREENER_CHOICE

Catalog No. 4465804
Encompass_Preferred
Change view
Click to view available options
Quantity:
Gene Reference (75 reactions), Inventoried
Mutant Allele (75 reactions), Inventoried
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-658-04 Mutant Allele (75 reactions), Inventoried
44-658-07 Gene Reference (75 reactions), Inventoried
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-658-04 Supplier Applied Biosystems™ Supplier No. 4465804
Add to Cart
Add to Cart

Powered by competitive allele-specific TaqMan™ PCR (castPCR™ Technology) to detect and measure somatic mutations in genes associated with cancer research.

The castPCR technology is highly specific and sensitive, and can detect rare amounts of mutated DNA in a sample that contains large amounts of normal, wild-type DNA. These assays are compatible with different sample types, such as cell lines, formalin-fixed paraffin-embedded (FFPE) tissue samples, and fresh frozen tissue samples.

Competitive, allele-specific TaqMan PCR utilizes an allele-specific primer for mutant allele detection that competes with an MGB blocker oligonucleotide to suppress the wild-type background.

  • Higher specificity: designed to block the wild-type and amplify only the mutant
  • Higher sensitivity: designed to detect somatic mutations down to 1 cancer cell in 1000 normal cells
  • Faster workflow: 3 hours from sample to result
  • Detect multiple types of mutations: detect single- and multiple-nucleotide mutations and insertion/deletions (indels)
  • Wide dynamic range and excellent PCR efficiency: assays demonstrate at least 4 logs of dynamic range and an average efficiency of 100% ± 10%
  • Fast, simple workflow: like other TaqMan Assays, typically require 3 hours from sample to results, with minimum hands-on time

Order Info

Expiration date 2 years from the date of manufacturer. Shipping Conditions: Ambient Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage 1 tube containing a 10X mix of unlabeled PCR primers, a TaqMan MGB probe, and an MGB oligonucleotide blocker.

Store at -15 to -25°C.

Detection Method Primer-probe
Form Frozen
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
Product Line TaqMan
Product Type Mutation Detection Assay
Quantity Mutant Allele (75 reactions), Inventoried
Shipping Condition Room Temperature
Fidelity (vs. Taq) 10 X
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.