Learn More
Applied Biosystems™ TaqMan™ OpenArray™ Genotyping Plate, Custom Format 60 QuantStudio™ 12K Flex
Description
Design your own PCR plate, building from custom assays and over 4.5 million predesigned genotyping assays, including 2,700 inventoried Drug Metabolizing Enzyme (DME) assays. The subsequent qPCR system workflow facilitates the validation/screening of up to 256 SNPs across >1,500 samples per day.
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Format | Custom Format 240 |
| Product Line | OpenArray, QuantStudio, TaqMan |
| Quantity | 10 pack |
Frequently Asked Questions (FAQs)
The format differences are due to the use of start and stop points. These are not used for GX arrays. In format 32, the 2 sample/subarray format is loaded mirrored. If we did this for GX, we would only end up being able to put assays in columns 1-4 of the subarray (5-6 contains the start points and 7-8 the stop points). The result would be just 16 assays. These are not made as the format was not deemed useful.
Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.
These instructions may be found on page 79 of the ProFlex PCR System User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0007697.pdf).
Reaction volume: 33 nL
Cover temperature: 70°C
94°C for 600 sec, ramp rate 1.3
50 cycles of:
99°C for 30 seconds, ramp rate 1.3
54°C for 21 seconds, ramp rate 1.1
57°C for 90 seconds, ramp rate 0.1
These instructions may be found on page 5 of the Dual Flat Block GeneAmp PCR System 9700 User Guide (https://tools.thermofisher.com/content/sfs/manuals/cms_065075.pdf).
93°C for 10 min, ramp rate 100%
50 cycles of:
95°C for 45 sec, 84% ramp rate
94°C for 13 sec, 100% ramp rate
53°C for 2 min 14 sec, 44% ramp rate
25°C for 2 min, 100% ramp rate
Optional: Add a post-PCR overnight hold as shown below:
4°C for 99 min 59 sec, 100% ramp rate
You need an OpenArray Plate Adaptor, Cat. No. 4472306. You do not need a compression pad. This part number works with both the GeneAmp PCR System 9700 and the ProFlex PCR System.
Please note that this part number does not have a product page on our website. To order, please call our customer service team at 800-955-6288, option 1, option 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.