Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TaqMan™ OpenArray™ RT PCR, Custom Format 56 QuantStudio™ 12K Flex

Catalog No. 4471120
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-711-20 10 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-711-20 Supplier LIFE TECHNOLOGIES Supplier No. 4471120
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Design your own plate, building from custom, inventoried, and pre-designed TaqMan® Gene expression assays. The OpenArray® Real-Time PCR System workflow enables one person to validate or screen up to 576 samples in one day without the use of robotics.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Primer-probe
Format Format 56
GC-Rich PCR Performance Low
PCR Method qPCR
Product Line OpenArray, QuantStudio, TaqMan
Product Type Gene Expression Assay
Quantity 10 pack
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.