Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Ribosomal RNA Control Reagents

Catalog No. 4308329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1,000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-083-29 1,000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-083-29 Supplier Applied Biosystems™ Supplier No. 4308329
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TaqMan™ Ribosomal RNA Control Reagents

Applied Biosystems™ TaqMan™ Ribosomal RNA Control Reagents are designed to detect the 18S ribosomal RNA (rRNA) gene. The TaqMan™ probe and primer sequences were designed following Rapid Assay Development Guidelines. The TaqMan™ probe and primer sequences are conserved among a diverse group of eukaryotes including human, rat, mouse, Xenopus, Saccharomyces, Giardia, and Arabidopsis. Ribosomal RNA levels provide an endogenous control for PCR quantitation studies.

  • Pre-designed primers and TaqMan™ probe eliminate assay design

  • Rapid Assay Development Guidelines can minimize optimization time

  • Separate components provide flexibility in assay set-up

    Comprehensive Guidelines Included

    Applied Biosystems™ has developed a comprehensive set of guidelines to ensure success when using Applied Biosystems™ Real-Time PCR reagents and instrumentation. These guidelines are simple and easy to follow. Many traditional variables, such as magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, greatly reducing assay development time.

    Normalization Built-In

    All Applied Biosystems™ Real-Time PCR reagent kits contain a passive internal reference to normalize non-PCR related fluorescence fluctuations. Normalizing with a passive internal reference minimizes well-to-well variability that can result from a variety of causes, such as pipetting errors and sample evaporation. The passive reference is also essential for accurate results when multiple probes (with different reporter dyes) are combined in a single tube.

    Notes on Use

    The TaqMan™ Ribosomal RNA Control Reagents can be used with TaqMan™ Universal PCR Master Mix or TaqMan™ PCR Core Reagents for polymerase chain reaction (PCR).

    The TaqMan™ Ribosomal RNA Control Reagents can be used with the TaqMan™ One-Step RT-PCR Master Mix Reagents, TaqMan™ EZ RT-PCR Core Reagents, or the TaqMan™ Gold RT-PCR Reagents for one-step reverse transcription polymerase chain reaction (RT-PCR).

    Notice to Purchaser

    Disclaimer of License: The TaqMan™ Ribosomal RNA Control Reagents are optimized for use in the polymerase chain reaction (PCR) covered by patents owned by Roche Molecular Systems, Inc. and F. Hoffmann-La Roche Ltd. No license under these patents to use the PCR Process is conveyed expressly or by implication to the purchaser by the purchase of these products. A license to use the PCR Process for certain research and development activities accompanies the purchase of certain Applied Biosystems™ reagents when used in conjunction with an authorized thermal cycler, or is available from Applied Biosystems™. Further information on purchasing licenses to practice the PCR Process may be obtained by contacting the Director of Licensing at Applied Biosystems™, 850 Lincoln Centre Dr., Foster City, California 94404 or Roche Molecular System, Inc., 1145 Atlantic Avenue, Alameda, California 94501.

    TaqMan™ probes are covered by US Patent 5,723, 591 and foreign counterparts and patents pending owned by Applied Biosystems™, and may be covered by US Patents 5,801,155 and 6,084,102 and foreign counterparts licensed to Applied Biosystems™.

    Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Control Primer-Probe Set(s) & Template(s)
    Control Template Human Control RNA
    Format Tube
    GC-Rich PCR Performance Low
    PCR Method 1-step RT-qPCR
    Reaction Speed Standard
    Gene Symbol 18S
    Internal Probe Modification TAMRA™ Quencher (3'), VIC™ (5')
    Label or Dye VIC
    Product Line TaqMan
    Product Type Real Time PCR Reagents
    Quantity 1,000 reactions
    Shipping Condition Dry Ice
    Species S. cerevisiae, Human, Rat, Xenopus, Giardia, Mouse, Arabidopsis
    Sufficient For 1000 Reactions
    Target 18S rRNA
    No. of Reactions 1000 Reactions
    Show More Show Less
    I am working on both mouse and human samples for gene expression studies. Do you have one endogenous control that I can use with both species?

    The 18S rRNA is highly conserved among eukaryotes. We have several configurations of assays for this gene: P/N 4331182, Assay ID: Hs99999901_s1; PN 4319413E, 4310893E, 4333760T, 4333760F, 4308329. You can use any of these for both human and mouse samples, as well as samples from any other eukaryotic species.

    What targets will the primer/probe set included in the TaqMan Ribosomal RNA Control Reagents kit (P/N 4308329) amplify?

    The TaqMan Ribosomal RNA Control Reagents kit (P/N 4308329) will amplify all eukaryotic 18S rRNA sequences. For more information on this kit, please refer to the TaqMan Ribosomal RNA Control Reagents Protocol.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.