Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ RNase P Control Reagents Kit

Catalog No. 4316844
Encompass_Preferred
Change view
Click to view available options
Quantity:
1,000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-168-44 1,000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-168-44 Supplier Applied Biosystems™ Supplier No. 4316844
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

TaqMan™ RNase P Control Reagents Kit

The RNase P gene is a single-copy gene that encodes the RNA moiety for the RNase P enzyme. Applied Biosystems™ TaqMan™ RNase P Control Reagents were designed with limiting primer concentrations to be used as the endogenous reference in multiplex reactions. Multiplex PCR is the use of more than one probe and primer pair in the same tube. This kit is designed to perform a 5' nuclease assay with TaqMan™ Universal PCR Master Mix using the provided human genomic DNA as template.

  • Pre-designed primers and TaqMan™ probe eliminate assay design

  • Easy-to-use primer and TaqMan™ probe mix minimizes assay set-up time

  • Designed with limiting primer concentrations to eliminate assay optimization

  • Rapid assay development guidelines can minimize optimization time

    Comprehensive Guidelines Included

    Applied Biosystems™ has developed a comprehensive set of guidelines to ensure success when using Applied Biosystems™ Real-Time PCR reagents and instrumentation. These guidelines are simple and easy to follow. Many traditional variables, such as magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, greatly reducing assay development time.

    Normalization Built-In

    All Applied Biosystems™ Real-Time PCR reagent kits contain a passive internal reference to normalize non-PCR related fluorescence fluctuations. Normalizing with a passive internal reference minimizes well-to-well variability that can result from a variety of causes, such as pipetting errors and sample evaporation. The passive reference is also essential for accurate results when multiple probes (with different reporter dyes) are combined in a single tube.

    Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Calibration Type Passive Internal Reference Standards
    Content And Storage Control Primer-Probe Set(s) & Template(s)
    Control Template Human Genomic DNA
    Format Tube
    GC-Rich PCR Performance Low
    PCR Method Multiplex qPCR
    Reaction Speed Standard
    Internal Probe Modification TAMRA™ Quencher (3'), VIC™ (5')
    Label or Dye VIC
    Product Line TaqMan
    Product Type Real Time PCR Reagents
    Quantity 1,000 reactions
    Shipping Condition Wet Ice
    Species Human
    Sufficient For 1000 Reactions
    Target RNase P
    No. of Reactions 1000 Reactions
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.