Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TARBP2 Mouse anti-Human, Clone: OTI2C12, liquid, TrueMAB™
SDP

Catalog No. 501681993
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1993 Supplier OriGene Supplier No. TA808239
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRBP is an RNA associated nuclear protein. It contains 3 DRBM (double stranded RNA-binding) domains and plays an important role by binding to HIV-1 TAR RNA located in the long terminal repeat (LTR).TRBP acts synergistic with viral TAT protein in HIV-1 expression activation. It binds preferentially to TAR and other double-stranded RNAs with G+C-rich characteristics. It is important in regulation of transcription from RNA polymerase II promoter andis widely expressed in most of the tissues.

Specifications

Antigen TARBP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TARBP2
Gene Accession No. Q15633
Gene Alias LOQS, TRBP, TRBP1, TRBP2
Gene Symbols TARBP2
Host Species Mouse
Immunogen Full length human recombit protein of human TARBP2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6895
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.