Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tartaric Acid Reagent 2, Reagents
Tartaric Acid Reagent 2, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CT10130020F
Quantity | 20 L |
---|
Chemical Identifiers
7732-18-5, 87-69-4 | |
18.02 | |
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.14 | |
Non-sterile | |
Tartaric Acid Reagent 2 |
7732-18-5, 87-69-4 | |
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
O | |
18.02 | |
Cubitainer | |
20 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction