Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tashiro's indicator solution, In ethanol, Solstice

Catalog No. 60090564 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
6 x 250 mL
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-564 6 x 1 L
60-016-37 250 mL
60-016-36 1 L
60-048-586 6 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-090-564 Supplier Solstice Supplier No. 360836X1L

In ethanol

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tashiro’s indicator solution
Name Note In ethanol
Boiling Point 78°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Grade Analytical Reagent
Packaging Glass Bottle
Density 0.79 g/cm3
Quantity 6 x 1 L
Flash Point 12°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.