Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TASK1 Potassium Channel Monoclonal Antibody (S374-48), QED Bioscience™
SDP

Catalog No. 502277455
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7455 Supplier Leinco Technologies Inc Supplier No. 11578100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TASK1 Potassium Channel Monoclonal antibody specifically detects TASK1 Potassium Channel in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen TASK1 Potassium Channel
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S374-48
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Gene KCNK3
Gene Accession No. O14649, O35111, O54912
Gene Alias acid-sensitive potassium channel protein TASK; acid-sensitive potassium channel protein TASK-1; cardiac potassium channel; cardiac two pore background K(+) channel; Ctbak; cTBAK 1; cTBAK-1; K2p3.1; KCNK3; KCNK9; LOW QUALITY PROTEIN: potassium channel subfamily K member 3; OAT1; Potassium channel subfamily K member 3; potassium channel subfamily K member 3; LOW QUALITY PROTEIN: potassium channel subfamily K member 3; potassium channel, subfamily K, member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; potassium two pore domain channel subfamily K member 3; PPH4; rTASK; TASK; TASK 1; TASK1; TASK-1; TASK-1 ion channel receptor; TASK-related potassium channel; TBAK1; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Host Species Mouse
Immunogen Fusion protein corresponding to aa 251-411 (cytoplasmic C-terminus) of rat TASK1. This sequence is 96
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16527, 29553, 3777
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.