Learn More
AdipoGen Taspine
SDP
Supplier:  AdipoGen AGCN20544M010
Encompass_Preferred
Chemical. CAS 602-07-3. Formula C20H19NO6. MW 369.4. Taspine is an alkaloid with multifunctional pharmacological properties, including apoptosis-inducing, anti-angiogenic, anticancer, cytotoxic, anti-inflammatory and wound-healing activities, thereby modulating multiple targets. Taspine is a potent acetylcholinesterase AChE inhibitor and wound healing promoter cicatrizant, by regulating the expression of keratinocyte growth factor KGF FGF-7 in early phases. Taspine has anticancer properties targeting DNA topoisomerase IB and was shown to be a dual topoisomerase inhibitor. Taspine is an apoptosis-inducing compound. It induces conformational activation of the pro-apoptotic proteins Bak and Bax, mitochondrial cytochrome c release and mitochondrial membrane permeabilization. It induced tumor apoptosis in vitro and in vivo. Taspine can inhibit tumor angiogenesis acting as a multi-target VEGFR-2 inhibitor.
Catalog No. 50-256-2477
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.