Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TAT-PDHPS1
GREENER_CHOICE

Catalog No. 78321
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-321 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-321 Supplier Tocris Bioscience™ Supplier No. 7832/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Peptide inhibitor of YAP signaling

TAT-PDHPS1 is a peptide inhibitor of Yes-associated protein (YAP) signaling. It comprises the endogenous peptide PDHPS1 and the cell-penetrating peptide sequence, TAT. PDHPS1 binds to protein phosphatase 2 phosphatase activator (PTPA), which activates protein phosphatase 2A (PP2A). This activation leads to the phosphorylation of YAP and the suppression of YAP-targeted genes. TAT-PDHPS1 inhibits the proliferation of ovarian cancer cells in vitro and ovarian tumor growth in a subcutaneous xenograft mouse model.

Specifications

Gene ID (Entrez) 10413
Purity 95%
Content And Storage Store at -20°C
Gene Alias 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog
Molecular Weight (g/mol) M.W. (Observed): 3744.32: 0.95
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.