Learn More
CPC Scientific H-Met-Ala-Glu-Pro-Arg-Gln-Glu-Phe-Glu-Val-Met-Glu-Asp-His-Ala-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SIGN016A
Encompass
This is the amino acids 1 to 16 fragment of the human Tau protein. Amyloid pathology is accompanied by the presence of various isoforms of this human protein in neurofibrillary tangles. This peptide was used as an immunogen to develop the anti-Tau antibody BR133ONE-LETTER SEQUENCE: MAEPRQEFEVMEDHAGMOLECULAR FORMULA: C78H118N22O28S2MOLECULAR WEIGHT:1876.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: Spillantini, M. et al. Proc. Natl. Acad. Sc. 87, 3947 (1990).
Catalog No. 50-211-0412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.