Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tau Synthetic Blocking Peptide, Human, Phospho S214, 500 μg

Catalog No. 502704395
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4395 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4395 Supplier Abcam Supplier No. AB19123500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic human Tau (phospho S214) peptide suitable for blocking applications.

  • Peptide Type: Synthetic
  • Species Reactivity: Human
  • Purity: >70%
  • Tested Applications: BL
  • Form: Lyophilized

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Tau (Phospho S214), Human
Shipping Condition Blue Ice
Substrate Properties Peptide-based Substrate
Content And Storage Store at -20°C.
Detection Location In-Blot Detection
Form Lyophilized
Label or Dye Unconjugated
Reactivity Human
Cross Reactivity Human
Quantity 500 μg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.