Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tauber’s Reagent, Reagents
Tauber’s Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CT0005004B
Quantity | 4 L |
---|
Chemical Identifiers
7732-18-5, 7664-38-2, 1310-73-2 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Light Yellow to Pale Orange | |
100°C | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0402 | |
Non-sterile | |
Tauber’s Reagent |
7732-18-5, 7664-38-2, 1310-73-2 | |
H2O | |
O | |
18.02 | |
Opaque HDPE | |
4 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction