Learn More
American Radiolabeled Chemicals Inc TAUROCHOLIC AC 14C 10UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 348910UC
Encompass_Preferred
Taurocholic acid-taurine-1,2-14C 10uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 515.7, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.