Learn More
Selleck Chemical LLC Tauroursodeoxycholic Acid (TUDCA) 25mg 14605-22-2
SDP
Supplier:  Selleck Chemical LLC S365425MG
Encompass_Preferred
Tauroursodeoxycholic acid (TUDCA) is the taurine conjugate of ursodeoxycholic acid (UDCA) and acts as a mitochondrial stabilizer and anti-apoptotic agent in several models of neurodegenerative diseases, including AD, Parkinson's diseases (PD), and Huntington's diseases (HD). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.