Learn More
Enzo Life Sciences Paclitaxel, (5mg), CAS Number: 33069-62-4
SDP
Supplier:  Enzo Life Sciences BMLT1040005
Encompass_Preferred
Ultra-pure; stabilizes microtubules. Antitumor agent. Stabilizes microtubules and lowers the critical concentration for tubulin polymerization. Reversibly binds to polymerized tubulin. Inhibits protein isoprenylation and induces apoptosis in PC-3 human prostate cancer cells. The checkpoint of mitotic spindle assembly, aberrant activation of cyclin-dependent kinases (CKDs) and the c-Jun N-terminal kinase/stress activated protein kinase (JNK/SAPK) are involved in Paclitaxel-induced apoptosis. Formula: C47H51NO14. MW: 853.9. CAS Number: 33069-62-4. MI: 14: 6982. Purity: ≥98% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO or methanol at 10mg/ml. Use/Stability: Store solutions at -20°C for up to 6 months. Long Term Storage: -20°C
Catalog No. NC9019658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.