Learn More
Description
Biological Activity: Antitumor agent; promotes and stabilizes tubulin polymerization, causing cell cycle arrest. Induces autocatalytic activation of caspase-10 in CCRF-HSB-2 cells, triggering apoptosis.
Storage: Store at 4°C.
Note: Inhibitors are guaranteed for 1 year from date of receipt.
Specifications
Specifications
| Formulation | C47H51NO14 |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 10 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 853.92g/mol |
| Solubility | Soluble to 100mM in DMSO, 25mM in ethanol |
| Content And Storage | 4°C |
| Target | Taxol |
| Color | White |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
