Learn More
ATCC Taylorella equigenitalis (type strain NCTC 11184) - Freeze-dried culture, ATCC 35865
Supplier: ATCC 35865
Taylorella equigenitalis (type strain NCTC 11184) is provided as a freeze-dried research-use-only culture derived from an equine cervical swab. The organism is microaerophilic and associated with contagious equine metritis; it is intended for laboratory research and must be handled under appropriate biosafety procedures.
- Freeze-dried vial format for long-term storage.
- Store at 2°C to 8°C.
- Culture on Trypticase Soy agar/broth or GC agar/broth for growth.
- Incubate at 37°C in 95% air, 5% CO2.
- Isolated from an equine cervical swab (England).
- Biosafety level 2 organism; handle with appropriate containment and procedures.
- Type strain designation NCTC 11184 (also referenced as 61717/77, CIP 7909).