Learn More
TARGETMOL CHEMICALS INC TAK-243 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1697410MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TAK-243 (MLN7243) is a small molecule inhibitor of ubiquitin-activating enzyme (UAE IC50=1 nM) which blocks ubiquitin conjugation disrupting monoubiquitin signaling as well as global protein ubiquitination. TAK-243 (MLN7243) induces endoplasmic reticulum (ER) stress abrogates NF-(kappa)B pathway activation and promotes apoptosiswith potential antineoplastic activity. purity: 98%
Catalog No. 50-258-1133
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.