Learn More
MilliporeSigma™ TB-fluor Staining Kit
Catalog No. M1090930001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 x 500mL
Catalog No. Quantity
M1090930001 6 x 500mL
1 options
Catalog No. M1090930001 Supplier MilliporeSigma™ Supplier No. 1.09093.0001
Identifies acid resistant mycobacteria
  • Used alongside Ziehl-Neelson staining (Tb-color modified)
  • Ready to use solutions
  • Phenol-free staining kit
  • Two fluorescent dyes, Rhodamine B – staining red - and Auramine O – staining yellowish-green
  • Ppenetration of the dyes into target structures is achieved through phenol substitute
  • Suppresses troublesome intrinsic fluorescence of specimen
Application:
  • Designed to be used for smears, paraffin sections
TRUSTED_SUSTAINABILITY
Specifications
Quantity 6 x 500mL
Packaging Fibre Carton