Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566916 Supplier Invitrogen™ Supplier No. PA566916
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APPAPSSTERE QSVRKSSTFP RTGYDSVKLY SPTSKALTRS DDVSVCSVSS LGTELSTTLS VSNEDILDLV VTSSSSAIVT LENDDDP Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.

Plays a role in the regulation of starvation-induced autophagosome formation (PubMed:22613832). Together with the TRAPPIII complex, regulates a constitutive trafficking step from peripheral recycling endosomes to the early Golgi, maintaining the cycling pool of ATG9 required for initiation of autophagy. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D14
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D14
Gene Accession No. Q9P2M4
Gene Alias 2810413P16Rik; AU043625; C86258; D5Ertd110e; Kiaa1322; mKIAA1322; TBC1 domain family member 14; TBC1 domain family, member 14; Tbc1d14
Gene Symbols TBC1D14
Host Species Rabbit
Immunogen Recombinant Human TBC1D14. Recombinant protein control fragment (Product #RP-107569).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.