Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560946 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560946 Supplier Invitrogen™ Supplier No. PA560946
  • $617.00
    $498.65 / Each of 1

    Save $118.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: FQETDKAWHG GCLALAELGR RGLLLPSRLV DVVAVILKAL TYDEKRGACS VGTNVRDAAC YVCWAFARAY EPQELKPFVT AISSALVIAA VFDRDINCRR AASAAFQENV GRQGTFPHGI DILTTADYFA VGN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tbcd
Gene Accession No. Q9BTW9
Gene Alias 2310057L06Rik; A030005L14Rik; Beta-tubulin cofactor D; KIAA0988; mKIAA0988; PP1096; SSD1; SSD-1; TBCD; TFCD; tubulin folding cofactor D; tubulin-folding cofactor D; Tubulin-specific chaperone D
Gene Symbols Tbcd
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBCD. Recombinant protein control fragment (Product #RP-102109).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.