Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBCEL Monoclonal Antibody (OTI2A9)
GREENER_CHOICE

Catalog No. PIMA525495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525495 Supplier Invitrogen™ Supplier No. MA525495
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBCEL Monoclonal Antibody for Western Blot, IHC (P)

Acts as a regulator of tubulin stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCEL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A9
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TBCEL
Gene Accession No. Q5QJ74
Gene Alias catastrophin; D330014K23; E130107N23Rik; El; E-like; leucine rich repeat containing 35; leucine-rich repeat-containing protein 35; Lrrc35; RGD1308880; Tbcel; tubulin folding cofactor E like; tubulin folding cofactor E-like; tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like
Gene Symbols TBCEL
Host Species Mouse
Immunogen Full length human recombinant protein of TBCEL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219899
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.