Learn More
Description
- Quality tested; demonstrates ≤1% release of DNase and RNase activity
Specifications
Specifications
| Chemical Name or Material | TBE Buffer 10X |
| Name Note | For polyacrylamide and agarose gel electrophoresis |
| Color | Colorless |
| CAS | 7732-18-5 |
| For Use With (Application) | Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Solution Type | Buffer |
| Synonym | Tris-borate-EDTA |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.