Learn More
Thermo Scientific nanoViper Fingertight Fitting Systems | | 1 EA
Supplier:  Thermo Scientific 6041.5294
Encompass
For Use With Application: Capillary LC Column to MS Inlet Connection, For Use With Equipment: Dionex UltiMate 3000 RSLC Nano System, Material: Shielded Fused Silica, Max Pressure: 1200 bar, Product Line: nanoViper, Type: Advanced Tubing, Length Metric: 1 m, Diameter Metric Inner: 50 m
Catalog No. NC2071530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.