Learn More
Selleck Chemical LLC TBK1/IKK-IN-1 5mg
SDP
Supplier:  Selleck Chemical LLC S89225MG
Encompass_Preferred
TBK1/IKK?-IN-1 (compound 1) is a dual inhibitor ofTANK-binding kinase 1 (TBK1) and I?B kinase-? (IKK?/IKK-i) with IC50 of 1.0 nM and 5.6 nM for TBK1 and IKK?, respectively. TBK1/IKK? inhibition enhances response to PD-1 blockade, which effectively predicts tumor response in vivo. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3437
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.