Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBL1X/TBL1XR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674388
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-388 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-388 Supplier Invitrogen™ Supplier No. PA19108
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is expected to recognize the human protein TBL1XR1 (NP_078941) as well as TBL1X. Variants (NP_005638.1; NP_001132938.1) encode isoform a (TBL1X) and variants (NP_001132939.1; NP_001132940.1 ) encode isoform b (TBL1X). This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroid receptors) complex along with histone deacetylase 3 protein. This gene is located adjacent to the ocular albinism gene and it is thought to be involved in the pathogenesis of the ocular albinism with late-onset sensorineural deafness phenotype. Four transcript variants encoding two different isoforms have been found for this gene. This gene is highly similar to the Y chromosome TBL1Y gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBL1X/TBL1XR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TBL1X
Gene Accession No. O60907, Q9BZK7
Gene Alias 5330429M20; 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; EBI; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1X; F-box-like/WD repeat-containing protein TBL1XR1; Ira1; MRD41; nuclear receptor co-repressor/HDAC3 complex subunit; nuclear receptor corepressor/HDAC3 complex subunit TBLR1; SMAP55; TBL1; TBL1-related protein 1; Tbl1x; Tbl1xr1; TBLR1; transducin (beta)-like 1 X-linked; transducin (beta)-like 1 X-linked receptor 1; transducin (beta)-like 1X-linked; transducin (beta)-like 1X-linked receptor 1; transducin beta-like 1X-related protein 1; Transducin beta-like protein 1X; transducin-beta-like protein 1, X-linked
Gene Symbols TBL1X, Tbl1xr1
Host Species Goat
Immunogen Synthetic peptide corresponding to the C terminus amino acids SASDGSVCVLDLRK (aa 564-577).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6907, 79718
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.