Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Precision Pump Tubing, C-Flex ULTRA, L/S 35; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0643435
Encompass
Applications: General-purpose lab uses requiring high performance and long life Characteristics: Exceptional tensile and tear strength, good biocompatibility, low spallation, reduced residue and low gas permeability, smooth surface, translucent
Catalog No. NC0925507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.