Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568922 Supplier Invitrogen™ Supplier No. PA568922
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 100%.

TBPL2 is a transcription factor required in complex with TAF3 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBPL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TBPL2
Gene Accession No. Q6SJ95
Gene Alias Gm348; TATA box binding protein like 2; TATA box binding protein-like 2; TATA box-binding protein-like protein 2; TATA box-binding protein-related factor 3; TATA-box binding protein like 2; TBP2; TBPL2; tbpl2 {ECO:0000250; TBP-like protein 2; TBP-related factor 3; TBP-related factor 3 (Trf3); TRF3; UniProtKB:P62380}
Gene Symbols TBPL2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse TBPL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227606
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.