Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBRG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575916 Supplier Invitrogen™ Supplier No. PA575916
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBRG4
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TBRG4
Gene Accession No. Q5M9G9, Q91YM4, Q969Z0
Gene Alias 2310042P22Rik; AA120735; AA408001; AI527316; cell cycle progression 2 protein; cell cycle progression protein 2; Cell cycle progression restoration protein 2; CPR2; FAST 4; FAST kinase domain-containing protein 4; FAST kinase domains 4; FASTKD4; H_TD2522F11.8; KIAA0948; mKIAA0948; Protein TBRG4; R74877; Tb-12; TBRG4; transforming growth factor beta regulated gene 4; transforming growth factor beta regulator 4
Gene Symbols TBRG4
Host Species Rabbit
Immunogen Recombinant full length Human TBRG4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21379, 360977, 9238
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.