Learn More
General Data Healthcare™ SHUR/Stain™ Manual Stainer Accessories
Shop All Triangle Biomedical Sciences Inc. (TBS) Products

Description
- Accessory
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | SHUR/Stain Manual Stainer |
| Product Type | Replacement Well, White |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.