Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ SHUR/Stain™ Manual Stainer Accessories
SDP

Catalog No. 22048007 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Product Type:
Digital Timer
Replacement Label Strip, for SS-12L
Replacement Label Strip, for SS-6L
Replacement Label Strip,for SS-3L
Replacement Well, Green
Replacement Well, White
Slide Rack
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-048-007 Replacement Well, Green
22-048-005 Slide Rack
22-048-006 Replacement Well, White
22-048-008 Digital Timer
22-048-010 Replacement Label Strip, for SS-6L
22-048-011 Replacement Label Strip, for SS-12L
22-048-009 Replacement Label Strip,for SS-3L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-048-007 Supplier General Data Healthcare™ Supplier No. SSWLG
Add to Cart
Add to Cart

Accessories for manual stainers (SS-3L, SS-6L, SS-12L)

  • Accessory

Specifications

Color Green
For Use With (Equipment) SHUR/Stain Manual Stainer
Product Type Replacement Well, Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.