Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBST (Tris-Buffered Saline + Tween 20), 20X Solution, Molecular Biology Grade, Hoefer™
SDP

Catalog No. 03500537
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-500-537 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-537 Supplier Hoefer, Inc Supplier No. GR1541
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dilute to 1X with high purity H20 for a solution consisting of 25mM Tris, pH 7.4, 3.0mM KCl, 140mM NaCl and 0.05% Tween™ 20

Use in Western detection

Specifications

Chemical Name or Material TBS with Tween (TBST)
Quantity 1 L
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym Tris-Buffered Saline + Tween™ 20
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.