Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam tBuBHQ, SERCA Inhibitor, >99% Purity, Solid, 500 mg

Catalog No. 502707722
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7722 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7722 Supplier Abcam Supplier No. AB120276500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure SERCA inhibitor for biological research.

  • Compound type: Sarco-endoplasmic reticulum Ca²±-ATPase (SERCA) inhibitor
  • Purity: >99%
  • Molecular weight: 222.32 Da
  • Form: Solid
  • Chemical name: 1,4-Benzenediol, 2,5-bis(1,1-dimethylethyl)
  • CAS number: 88-58-4
  • Storage: Store at Ambient

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Sarco-endoplasmic Reticulum Ca2+-ATPase (SERCA)
Shipping Condition Ambient
Form Solid
Buffer 100% 1,4-Benzenediol, 2,5-bis(1,1-dimethylethyl)
Content And Storage Store at ambient temperature.
Formulation 100% 1,4-Benzenediol, 2,5-bis(1,1-dimethylethyl)
Quantity 500 mg
For Use With (Application) Sarco-endoplasmic reticulum Ca2+-ATPase (SERCA) inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.