Learn More
Abcam tBuBHQ, SERCA Inhibitor, Solid, Purity >99%, 100 mg
Description
- Molecular Weight: 222.32 Da
- Purity: >99%
- Biological Activity: Sarco-endoplasmic reticulum Ca²±-ATPase (SERCA) inhibitor
- Chemical Name: 1,4-Benzenediol, 2,5-bis(1,1-dimethylethyl)
- Physical Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Sarco-endoplasmic reticulum Ca²±-ATPase (SERCA) |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Quantity | 100 mg |
| For Use With (Application) | Sarco-endoplasmic reticulum Ca²+-ATPase (SERCA) inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.