Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX19 Monoclonal Antibody (CL6251)
GREENER_CHOICE

Catalog No. PIMA531431 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531431 Supplier Invitrogen™ Supplier No. MA531431
  • $690.00
    $518.65 / Each of 1

    Save $171.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Immunogen sequence: EVHASTPGAF LLGNPAVTSP PSVLSTQAPT SAGVEVLGEP SLTSIAVSTW TAVASHPFAG WGGPGAGGHH SPSSLDG Highest antigen sequence indentity to the following orthologs: Rat - 32%, Mouse - 82%.

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene were found in patients with isolated deficiency of pituitary POMC-derived ACTH, suggesting an essential role for this gene in differentiation of the pituitary POMC lineage. ACTH deficiency is characterized by adrenal insufficiency symptoms such as weight loss, lack of appetite (anorexia), weakness, nausea, vomiting, and low blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX19
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL6251
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tbx19
Gene Accession No. O60806
Gene Alias AU019250; D1Ertd754e; dJ747L4.1; si:ch211-155d24.2; T-box 19; T-box factor, pituitary; T-box protein 19; T-box transcription factor 19; T-box transcription factor TBX19; T-box transcription factor TBX19; T-box-containing protein TBXT; TBS 19; TBS19; TBX19; tbx-19; Tpit; Tpit; Tbx19
Gene Symbols Tbx19
Host Species Mouse
Immunogen Recombinant protein fragment of Human TBX19 (Product #RP-108604)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9095
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.