Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 424800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-480-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-480-0 Supplier Invitrogen™ Supplier No. 424800
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

42-4800 has been successfully used in immunohistochemistry analysis of Tbx3 in mouse E16.5 frozen lung sections.

T-box 3 (TBX3) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. The Tbx3 protein is a transcriptional repressor that binds the canonical Brachychury bindig sites, and is thought to play a role in the anterior/posterior axis of the tetrapod forelimb. Tbx3 is required for normal mammary development and is also implicated in tumor development. Tbx3 expression also increases during osteoblast differentiation, and may function as a determinant of osteoblast cell numbers. Mutations in T-box 3 cause ulnar-mammary syndrome (MIM 181450), affecting limb, apocrine gland, tooth, hair, and genital development. Alternative splicing results in three transcript variants encoding different isoforms; however, the nature of one full length variant has not been determined. Subsets of human breast cancer cell lines overexpress TBX36. in vitro, Tbx3 overexpression in mouse embryo fibroblasts leads to immortalization of cells. Truncated forms of Tbx3 are increased in plasma samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Tbx3
Gene Accession No. O15119, P70324, Q7TST9
Gene Alias bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; TBX3; TBX3-ISO; UMS; XHL
Gene Symbols Tbx3
Host Species Rabbit
Immunogen Synthetic peptide derived from an internal region of human T-box 3, which differs from mouse and rat by one conservative and one non-conservative amino acid.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21386, 353305, 6926
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.