Learn More
Cayman Chemical TC 2559fumarate 25mg
SDP
Supplier:  Cayman Chemical 2560025 MG
Encompass_Preferred
A naphthoquinone with diverse biological activities inhibits U-46619-induced rabbit platelet aggregation in vitro (IC50 0.58 M) inhibits the conversion of arachidonic acid to thromboxane B2 in rabbit platelets inhibits human platelet aggregation induced by ADP collagen epinephrine and A23187 in vitro (IC50s 3.21 15.69 8.44 and 44.2 M respectively) increases tail bleeding time and exhibits a protective effect against pulmonary thrombosis in mice at 50 and 100 mg/kg inhibits CD45 (IC50 0.29 M) inhibits ovalbumin-induced footpad swelling in a mouse model of delayed-type hypersensitivity at 3 mg/kg is active against C. albicans A. niger B. subtilis S. aureus E. coli and P. aeruginosa (MICs 12.5-50 g/ml)
Catalog No. 50-293-6732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.