Learn More
Cole-Parmer Digi-Sense Temp-100 Dual-Input Thermocouple Thermometer
Supplier:  Cole-Parmer 9142803
Encompass_Preferred
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Select from the three most popular thermocouple types to meet all your basic temperature measurement needs. Digi-Sense's exclusive "3-WAY HANDS-FREE" capability allows "hands-free" use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meters come standard with the integral stand; the "hands-free" option adds magnets and strap (order separately).
Catalog No. 50-196-1600
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.