Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC JL 37
GREENER_CHOICE

Catalog No. p-7116101
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-251-0 10 mg
61-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-251-0 Supplier Tocris Bioscience™ Supplier No. 6125/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TYK2 inhibitor

TC JL 37 is a potent inhibitor of tyrosine kinase 2 (TYK2, Ki = 1.6 nM). Shows some selectivity for TYK2 over JAK1/2/3. Inhibits IFNγ production in vivo in mice. Orally bioavailable.

Specifications

Chemical Name or Material 2-Chloro-4-cyano-6-fluoro-N-[2-[[[(1R,2R)-2-fluorocyclopropyl]carbonyl]amino]-4-pyridinyl]benzamide
CAS 1258294-34-6
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C17H11ClF2N4O2
Formula Weight Observed MW: 376.74
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.