Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ TC Mayo Harrington/Metzenbaum Dissecting Scissors

Catalog No. 21124491 Shop All Surgical Design Products
Change view
Click to view available options
Includes:
Curved Blades, TC Inserts
Straight Blades, TC Inserts
Length (English):
5.75 in.
7 in.
9 in.
12 in.
16 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Length (English)
21-124-491 Curved Blades, TC Inserts 7 in.
21-124-480 Straight Blades, TC Inserts 9 in.
21-124-481 Straight Blades, TC Inserts 12 in.
21-124-482 Straight Blades, TC Inserts 16 in.
21-124-484 Curved Blades, TC Inserts 9 in.
21-124-485 Curved Blades, TC Inserts 12 in.
21-124-486 Curved Blades, TC Inserts 16 in.
21-124-488 Straight Blades, TC Inserts 5.75 in.
21-124-489 Curved Blades, TC Inserts 5.75 in.
21-124-490 Straight Blades, TC Inserts 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-124-491 Supplier Surgical Design™ Supplier No. 129TC
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dissecting scissors combine strength and precision for controlled tissue dissection and cutting.

  • Constructed from medical-grade stainless steel with tungsten carbide (TC) inserts for exceptional cutting performance and edge longevity
  • Available in 9-in., 12-in., and 16-in. lengths to accommodate various depths
  • Offered in both Straight and Curved tip configurations for surface and deep-tissue dissection
  • Mayo-Harrington design provides extra leverage and strength for cutting dense fascia and tissue
  • Sharp, precision-ground blades ensure smooth, clean cuts with minimal resistance
  • Gold-plated handles identify TC inserts and add visual contrast for professional handling
  • Ergonomic ring handles enhance comfort and stability during prolonged use
  • Ideal for general surgery, orthopedic, and soft-tissue dissection
  • Commonly used in pathology, autopsy, and veterinary surgical work
  • Suitable for academic, research, and training environments requiring precision and durability

Specifications

Product Type Dissecting Scissors
Material Stainless Steel with Tungsten Carbide Inserts
For Use With (Application) Dissection Related to: Pathology, Autopsy, Histology, Medical, Dental and Veterinary Applications
Includes Curved Blades, TC Inserts
Length (English) 7 in.
Length (Metric) 17.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.