Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC Mps1 12

Catalog No. 475010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-501-0 10 mg
47-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-501-0 Supplier Tocris Bioscience™ Supplier No. 4750/10
This item is not returnable. View return policy

Potent and selective Mps1 kinase inhibitor

TC Mps1 12 is a potent and selective monopolar spindle 1 (Mps1) inhibitor (IC50 = 6.4 nM). Selective for Mps1 over a panel of 95 kinases including JNK. Inhibits A549 lung carcinoma cell proliferation and attenuates A549 cell xenograft growth in mice. Orally active.

Specifications

Purity >99%
Quantity 10 mg
Product Type TC Mps1 12
Molecular Weight (g/mol) 324.38
Inhibitors TC Mps1 12

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.