Learn More
Eurofins Discoverx TC-O 9311, 10 mg. (92-1232)
Supplier:  Eurofins Discoverx 921232
Encompass
TC-O 9311 is a synthetic small molecule and potent agonist of the G protein-coupled receptor 139 (GPR139, EC50=39 nM). Lacks activity against a selected group of 90 other diverse targets. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.