Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TCBS Cholera Medium (Dehydrated)

Catalog No. OXCM0333B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0333B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0333B Supplier Thermo Scientific™ Supplier No. CM0333B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate pathogenic Vibrio species with this medium for highly reproducible results.

Selectively isolate pathogenic Vibrio species with Thermo Scientific™ Oxoid™ TCBS (Thiosulfate Citrate Bile Salts Sucrose) Cholera Medium (Dehydrated). The medium has shown to be the most satisfactory and reproducible formula regardless of the complex composition1. Unlike Lauryl Sulphate Tellurite Agar it does not require further additives or aseptic additions of blood. It also possesses superior growth characteristics for Vibrio species, compared with tellurite media. While inhibiting non-vibrios, it promotes rapid growth of pathogenic vibrios after overnight incubation at 35°C.

Vibrio is a comma-shaped Gram-negative bacteria. Vibrios are aquatic microorganisms, some species of which cause serious diseases in humans and other animals. Three species of Vibrio are of significance to humans: Vibrio cholerae is the cause of cholera, Vibrio parahemolyticus and V. vulnificus both act as agents of acute enteritis, or bacterial diarrhea.

Use TCBS Cholera Medium for selective isolation of pathogenic vibrios.

  • Produces reproducible results.
  • No need for additives or aseptic additions of blood.
  • Promotes rapid growth of pathogenic vibrios.

The TCBS medium conforms to the formulation of Kobayashi et al., except that it contains specially processed ox bile, free from the defects noted by Nakanishi and Kobayashi2.

The TCBS medium is suitable for growth of Vibrio cholerae, Vibrio parahemolyticus, and most other Vibrios3. Most Enterobacteriaceae encountered in feces are totally suppressed for at least 24 hours. Slight growth of Proteus species and Enterococcus fecalis may occur but the colonies are easily distinguished from Vibrio colonies.

 Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, ISO, NMKL.
Description Cholera Medium, TCBS
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 5.7L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.