Specifications
| Content And Storage | Store TcBuster-M at ≤ -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles by making single use aliquots at your desired volume. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 100 μg |
| Regulatory Status | GMP |